Learn More
Med Vet International ZipCombo Centrifuge with 6-Place Microtube Rotor, J0501ZC6, Each
Catalog No. 5000585084
Encompass_Preferred
ZipCombo Centrifuge. The JorVet ZipCombo is a multi-function micro-centrifuge available with a choice of rotors to meet all your micro-centrifugation needs in one unit. The flat hematocrit rotor spins shorter capillary tubes and eliminates angled separation lines, clay blow-outs, and broken tubes. The six place microtube rotor spins 1.5 to 2.0ml tubes for serum and urine separations and comes with adapters for other sized tubes (0.2ml - 1.5ml). Designed with robust construction for long life, the Zip Combo is well-suited to accomplish blood and urine separations, and microfiltrations. The small footprint makes the JorVet ZipCombo centrifuge the perfect addition to any clinic or lab. Features: - Interchangeable rotors - Clear lid for using tachometer - Digital LCD display of speed and time; lid open/close - Beep signals end of cycle - Suction-cupped feet to prevent slipping - Allen wrench included for changing rotors
Catalog No. 50-005-85084 Supplier Med Vet International Supplier No. JOR501ZC6
ZipCombo Centrifuge. The JorVet ZipCombo is a multi-function micro-centrifuge available with a choice of rotors to meet all your micro-centrifugation needs in one unit. The flat hematocrit rotor spins shorter capillary tubes and eliminates angled separation lines, clay blow-outs, and broken tubes. The six place microtube rotor spins 1.5 to 2.0ml tubes for serum and urine separations and comes with adapters for other sized tubes (0.2ml - 1.5ml). Designed with robust construction for long life, the Zip Combo is well-suited to accomplish blood and urine separations, and microfiltrations. The small footprint makes the JorVet ZipCombo centrifuge the perfect addition to any clinic or lab. Features: - Interchangeable rotors - Clear lid for using tachometer - Digital LCD display of speed and time; lid open/close - Beep signals end of cycle - Suction-cupped feet to prevent slipping - Allen wrench included for changing rotors