Learn More
AUTOMATIONDIRECT COM INC ZIPLINK STEPPER CABLE
Catalog No. NC3066574
Encompass
ZIPLink stepper cable, 6P4C RJ11 to 6-pin RJ12, RS-232, 6.5ft/2mcable length. For use with SureStep drives.
Catalog No. NC3066574 Supplier AUTOMATIONDIRECT COM INC Supplier No. STP232RJ12CBL2
ZIPLink stepper cable, 6P4C RJ11 to 6-pin RJ12, RS-232, 6.5ft/2mcable length. For use with SureStep drives.