Learn More
AUTOMATIONDIRECT COM INC ZIPLINK STPPR CBL 6P4C RJ11
Catalog No. NC3066616
Encompass
ZIPLink stepper cable, 6P4C RJ11 to 15-pin D-sub HD15 male,shielded, twisted pair, RS-232, 6.5ft/2m cable length. For use withSureStep drives.
Catalog No. NC3066616 Supplier AUTOMATIONDIRECT COM INC Supplier No. STP232HD15CBL2
ZIPLink stepper cable, 6P4C RJ11 to 15-pin D-sub HD15 male,shielded, twisted pair, RS-232, 6.5ft/2m cable length. For use withSureStep drives.