Learn More
Cole-Parmer Zip-lip low-density polyethylene bag with white labeling area, 5" x 8"
Catalog No. NC9411224 Shop All Cole-Parmer Products
Encompass_Preferred
Low-density polyethylene. Zip-lip seal allows dustproof, moisture-proof storage. All bags are single track, and have a white block marking area. Mark with pen, pencil, rubber stamp, or Cole-Parmer Lab Marker. Not for use with liquids.
Catalog No. NC9411224 Supplier Cole-Parmer Supplier No. 6010240
Low-density polyethylene. Zip-lip seal allows dustproof, moisture-proof storage. All bags are single track, and have a white block marking area. Mark with pen, pencil, rubber stamp, or Cole-Parmer Lab Marker. Not for use with liquids.