Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CritiCore Protective Wear Zipper Front Reusable Cleanroom Frock

Catalog No. 18111266
Change view
Click to view available options
Size:
2X-Small
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
5X-Large
6X-Large
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-111-266 -
18-111-258 2X-Small
18-111-259 X-Small
18-111-260 Small
18-111-261 Medium
18-111-262 Large
18-111-263 X-Large
18-111-264 2X-Large
18-111-265 3X-Large
18-111-267 5X-Large
18-111-268 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 18-111-266 Supplier CritiCore Protective Wear Supplier No. 12F23165N180014XLWHT
Add to Cart
Add to Cart
This item is not returnable. View return policy

Reusable frocks with coil zipper enclosure are suitable for a variety of cleanroom applications including pharmaceutical, biotechnology, and medical device manufacturing.

  • Fluid and bacteria resistant Integrity 1800™ high performance carbon striped fabric
  • Mandarin collar with two snaps at top
  • Wide-pull zipper with a single snap at bottom
  • Cuff Sleeves
  • Laser-cut, pre-serged seams
  • Silicon-free
  • ESD carbon stripes provide electrostatic dissipation

Specifications

Material Polyester
Color White
Disposable Reusable
Sterility Non-sterile
Closure Type Zipper with Snaps
Cuff Style Knit
Latex Free Yes
Length (Metric) 55.88 cm
Construction Laser-cut, pre-serged seams, silicon-free
Length (English) 22 in.
Product Type Controlled Environments Frock
Seam Type Pre-serged
For Use With (Application) Pharmaceutical, Electronics Assembly, Critical Environments
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.