Learn More
Cellucap SMS Coveralls, Open Wrists and Ankles
Catalog No. 19133810 Shop All Cellucap Products
Change view
Click to view available options
Size:
Small
4X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-133-810 4X-Large
19-168-320A Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-133-810 Supplier Cellucap Supplier No. 55802XXXX
Add to Cart
Add to Cart
This item is not returnable. View return policy
Soft and comfortable non-woven coveralls have two layers of spunbonded polypropylene enhanced with an inner layer of meltblown for extra protection.
  • Style 5502 SMS Coveralls
  • Similar to spunbond poylpropylene with soft and comfortable non-woven fabric
  • White, open wrists and ankles
  • Two layers of spunbonded polypropylene enhanced with an inner layer of meltblown for extra protection
  • Ideal for dusty areas and also provides splash protection
Specifications
Product Type General Purpose Coveralls
Size 4X-Large
Gender Unisex
Material SMS Fabric
Color White
Disposable Single-use
Ankle Style Open
Cuff Style Open
Hood Type Unhooded
Closure Type Front Zipper
Sterility Non-sterile
Resistance Dust
Description 3-ply protection, Spunbond Polypropylene
Model 5502