Learn More
Medchemexpress LLC Ziprasidone amino acid | 1159977-64-6 | MFCD00072662 | 99.5% | C21H23ClN4O2S | 10MG
SDP
Catalog No. 5000260675
Encompass_Preferred
Ziprasidone amino acid (Ziprasidone Impurity C) is an analytically characterized impurity of ziprasidone supplied as a high-purity solid reference standard for impurity profiling, method development, and quality control. It is intended for research and analytical use to support identification and quantification of open-ring ziprasidone-related impurities.
  • High purity suitable for analytical use.
  • Identified impurity for method development and validation.
  • Confirmed molecular formula C21H23ClN4O2S for reference.
  • Provided as a small, convenient mass for laboratory testing.
  • Includes structural identifiers for spectral and chromatographic correlation.
Catalog No. 50-002-60675 Supplier Medchemexpress LLC Supplier No. HY13125510MG
Add to Cart
Add to Cart
Ziprasidone amino acid (Ziprasidone Impurity C) is an analytically characterized impurity of ziprasidone supplied as a high-purity solid reference standard for impurity profiling, method development, and quality control. It is intended for research and analytical use to support identification and quantification of open-ring ziprasidone-related impurities.
  • High purity suitable for analytical use.
  • Identified impurity for method development and validation.
  • Confirmed molecular formula C21H23ClN4O2S for reference.
  • Provided as a small, convenient mass for laboratory testing.
  • Includes structural identifiers for spectral and chromatographic correlation.