Learn More
Medchemexpress LLC Ziprasidone-d8 | 1126745-58-1 | C21H13D8ClN4OS | 1 MG
SDP
Supplier:  Medchemexpress LLC HY14542S1MG
Encompass_Preferred
Ziprasidone-d8 is a deuterium-labeled compound that acts as a combined 5-HT (serotonin) and dopamine receptor antagonist. It exhibits potent antipsychotic activity and can be utilized as a tracer or internal standard in various analytical methods such as NMR, GC-MS, or LC-MS.
  • Deuterium labeled Ziprasidone
  • Functions as a 5-HT and dopamine receptor antagonist
  • Exhibits potent antipsychotic activity
  • Suitable for use as a tracer
  • Can be used as an internal standard for quantitative analysis
Catalog No. 50-003-56428