Learn More
Selleck Chemical LLC Ziprasidone HCl 10mg 122883-93-6 CP-88059
SDP
Catalog No. 501361259
Encompass_Preferred
Ziprasidone HCl (CP-88059) is a novel and potent dopamine and serotonin (5-HT) receptor antagonist, used in the treatment of schizophrenia and bipolar disorder. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1259 Supplier Selleck Chemical LLC Supplier No. S144410MG
Ziprasidone HCl (CP-88059) is a novel and potent dopamine and serotonin (5-HT) receptor antagonist, used in the treatment of schizophrenia and bipolar disorder. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.