Learn More
Apexbio Technology LLC Ziprasidone HCl 122883-93-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000093794
Encompass_Preferred
Ziprasidone hydrochloride (CAS 122883-93-6) is a small molecule classified as an atypical antipsychotic that exerts its pharmacological effects primarily through antagonism of serotonin 5-HT2A and dopamine D2 receptors It demonstrates high binding affinity for the 5-HT2A receptor with a 5-HT2A/D2 affinity ratio greater than those observed in other atypical antipsychotics (pKi for 5-HT2A 9 38) This dual antagonistic activity modulates neurotransmitter pathways implicated in neuropsychiatric disorders Ziprasidone HCl is frequently utilized in research to investigate serotonergic and dopaminergic mechanisms relevant to schizophrenia mood disorders and behavioral pharmacology
Catalog No. 50-000-93794 Supplier Apexbio Technology LLC Supplier No. A53505.1
Ziprasidone hydrochloride (CAS 122883-93-6) is a small molecule classified as an atypical antipsychotic that exerts its pharmacological effects primarily through antagonism of serotonin 5-HT2A and dopamine D2 receptors It demonstrates high binding affinity for the 5-HT2A receptor with a 5-HT2A/D2 affinity ratio greater than those observed in other atypical antipsychotics (pKi for 5-HT2A 9 38) This dual antagonistic activity modulates neurotransmitter pathways implicated in neuropsychiatric disorders Ziprasidone HCl is frequently utilized in research to investigate serotonergic and dopaminergic mechanisms relevant to schizophrenia mood disorders and behavioral pharmacology