Learn More
Medchemexpress LLC Ziprasidone hydrochloride | 122883-93-6 | 99.7% | C21H22Cl2N4OS | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14542A10MM/1ML
Encompass_Preferred
This product is an orally active compound that functions as a combined 5-HT and dopamine receptor antagonist. It exhibits high affinities for Rat D2, 5-HT2A, and 5-HT1A receptors.
  • Receptor antagonist activity.
  • High affinity for certain dopamine and 5-HT receptors.
  • Demonstrated to block wild-type hERG current in HEK-293 cells in a concentration-dependent manner.
  • In animal studies, it resulted in reduced weight gain, lower physical activity, higher resting energy expenditure, and increased capacity for thermogenesis when exposed to cold.
Catalog No. 50-003-55809