Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ziprasidone hydrochloride | 122883-93-6 | 99.7% | C21H22Cl2N4OS | 100 MG
SDP

Catalog No. 5000356256
Encompass_Preferred

Ziprasidone hydrochloride is an orally active combined 5-HT and dopamine receptor antagonist. It demonstrates high affinities for several serotonin and dopamine receptors, including Rat D2, 5-HT2A, and 5-HT1A.

  • Acts as an orally active combined 5-HT and dopamine receptor antagonist
  • Shows affinities for Rat D2 (Ki=4.8 nM), 5-HT2A (Ki=0.42 nM), and 5-HT1A (Ki=3.4 nM) receptors
  • Blocks wild-type hERG current in HEK-293 cells with an IC50 of 120 nm
  • In vivo studies suggest effects on weight loss, physical activity, energy expenditure, and thermogenesis

Catalog No. 50-003-56256 Supplier Medchemexpress LLC Supplier No. HY14542A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ziprasidone hydrochloride is an orally active combined 5-HT and dopamine receptor antagonist. It demonstrates high affinities for several serotonin and dopamine receptors, including Rat D2, 5-HT2A, and 5-HT1A.

  • Acts as an orally active combined 5-HT and dopamine receptor antagonist
  • Shows affinities for Rat D2 (Ki=4.8 nM), 5-HT2A (Ki=0.42 nM), and 5-HT1A (Ki=3.4 nM) receptors
  • Blocks wild-type hERG current in HEK-293 cells with an IC50 of 120 nm
  • In vivo studies suggest effects on weight loss, physical activity, energy expenditure, and thermogenesis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.