Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ziprasidone 98.0+%, TCI America™
SDP

Catalog No. Z0034100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z0034100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Z0034100MG Supplier TCI America Supplier No. Z0034100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 146939-27-7
Molecular Formula C21H21ClN4OS
Molecular Weight (g/mol) 412.936
MDL Number MFCD00866661
InChI Key MVWVFYHBGMAFLY-UHFFFAOYSA-N
PubChem CID 60854
ChEBI CHEBI:10119
IUPAC Name 5-[2-[4-(1,2-benzothiazol-3-yl)piperazin-1-yl]ethyl]-6-chloro-1,3-dihydroindol-2-one
SMILES C1CN(CCN1CCC2=C(C=C3C(=C2)CC(=O)N3)Cl)C4=NSC5=CC=CC=C54

Specifications

Melting Point 222°C
Quantity 100 mg
Formula Weight 412.94
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Ziprasidone
RTECSNumber NM3241000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.