Learn More
Cayman Chemical Ziprasidone (hydrochloride hydrate), Purity: ≥98%, CAS: 138982-67-9, 50 mg
SDP
Catalog No. 502969434
Encompass_Preferred
An atypical antipsychotic; an agonist of 5-HT1A receptors (EC50 = 36.31 nM for inhibition of forskolin-induced adenylate cyclase activity in HeLa cells expressing human receptors), as well as an inverse agonist of 5-HT1D receptors (IC50 = 2.69 nM) and a partial agonist of 5-HT1B receptors (EC50 = 6.17 nM) in [35S]GTPγS binding assays; an antagonist at 5-HT2A and dopamine D2 receptors (Kis = 1.15 and 1.29 nM, respectively) in cell-based assays; inhibits d-amphetamine-induced hyperactivity and apomorphine-induced stereotypy in rats (ID50s = 1.53 and 2.43 mg/kg, respectively)
Catalog No. 50-296-9434 Supplier Cayman Chemical Supplier No. 1503150 MG
An atypical antipsychotic; an agonist of 5-HT1A receptors (EC50 = 36.31 nM for inhibition of forskolin-induced adenylate cyclase activity in HeLa cells expressing human receptors), as well as an inverse agonist of 5-HT1D receptors (IC50 = 2.69 nM) and a partial agonist of 5-HT1B receptors (EC50 = 6.17 nM) in [35S]GTPγS binding assays; an antagonist at 5-HT2A and dopamine D2 receptors (Kis = 1.15 and 1.29 nM, respectively) in cell-based assays; inhibits d-amphetamine-induced hyperactivity and apomorphine-induced stereotypy in rats (ID50s = 1.53 and 2.43 mg/kg, respectively)