Learn More
Cayman Chemical ZIptIDtrIfluoROCTAte sal 10mg
SDP
Catalog No. 502918175
Encompass_Preferred
A peptide substrate for various serine/threonine protein kinases; has been used to evaluate CamKII activity for inhibitor development
Catalog No. 50-291-8175 Supplier Cayman Chemical Supplier No. 3746610 MG
A peptide substrate for various serine/threonine protein kinases; has been used to evaluate CamKII activity for inhibitor development