Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ ZipTip™ Pipet Tips
GSA_VA

Catalog No. ZTC04S096 Shop All MilliporeSigma Products
Change view
Click to view available options
For Use With (Application):
C18 Media
C4 Media
Micro Bed of C18 Media
Strong Cation Exchange Media
Quantity:
8/Pk.
96/Pk.
960/Pk.
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Quantity
ZTC04S096 C4 Media 96/Pk.
ZTC18S096 C18 Media 96/Pk.
ZTC04S008 C4 Media 8/Pk.
ZTSCXS096 Strong Cation Exchange Media 96/Pk.
ZTSCXS008 Strong Cation Exchange Media 8/Pk.
ZTC18S960 C18 Media 960/Pk.
ZTC18M960 Micro Bed of C18 Media 960/Pk.
ZTC18M096 Micro Bed of C18 Media 96/Pk.
ZTC18M008 Micro Bed of C18 Media 8/Pk.
ZTC18S008 C18 Media 8/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. ZTC04S096 Supplier MilliporeSigma™ Supplier No. ZTC04S096
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Sample preparation and precise liquid handling in one device

A 0.6μL bed of C18, C4, or SCX (strong cation exchange) media is fixed at the end of a 10μL pipet tip such that there is no dead volume; concentrated, purified samples are eluted in 1 to 4μL. For applications requiring smaller elution volumes (e.g., <1μL), ZipTip μ-C18 tips feature a microbed format containing 0.2μL of media.

  • Concentrate and purify 1–100μL biological samples
  • Ideal for purifying and concentrating femtomoles to picomoles of peptides, proteins, or oligonucleotides in minutes prior to mass spectrometry, HPLC, capillary electrophoresis, and other analytical techniques

Order Info

Available in two packaging configurations: resealable trays (packs of 8 and 24 tips) and 96-place tip racks (packs of 96 and packs of 960 tips)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 10 μL
Volume (Metric) Dispensing 10 μL
Sterility Non-sterile
Length (English) 1.25 in.
Length (Metric) 3.1cm
For Use With (Application) C4 Media
Tip Style Media-Fixed
Quantity 96/Pk.
Product Type Micro-SPE Pipette Tip, C4 Resin
Shipping Condition Ambient
For Use With (Equipment) Universal Fit
Product Line ZipTip
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.