Learn More
DSI LLC ZIRCHROM - WCX WEAK CATION-EX
Supplier:  DSI LLC ZR041021
Encompass_Preferred
ZirChrom-WCX. Weak Cation-exchange. 300 Angstrom. Useful for protein chromatography in the cation-exchange mode. Stable over a pH range of 1-10.
Catalog No. 50-123-091