Learn More
DSI LLC ZIRCHROM - WCX WEAK CATION-EX
Supplier: DSI LLC ZR041046
ZirChrom-WCX. Weak Cation-exchange. 300 Angstrom. Useful for protein chromatography in the cation-exchange mode. Stable over a pH range of 1-10.