Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Zirconium acetate solution

Catalog No. 413801500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-380-1500ML 500 mL
41-380-12L 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 41-380-1500ML Supplier Sigma Aldrich Supplier No. 413801500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Zirconium acetate solution is a metallic salt solution that can be used for a variety of applications such as: a zirconium based precursor for the development of zirconia coatingsa stabilizer for colloidal solutions, controlled growth of ice crystals, water proofing, and flame retardant treatments

Specifications

CAS 7585-20-8
Density 1.279 g/mL (at 25°C)
Quantity 500 mL
Synonym Acetic acid, zirconium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.