Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zirconium boride, 99.5% (metals basis excluding Hf), Thermo Scientific Chemicals

Catalog No. AA4703030
Click to view available options
Quantity:
50 g
250 g

Catalog No. AA4703030

Supplier: Thermo Scientific Chemicals 04703030

Only null left
Add to Cart

Zirconium Boride is used as an aerospace refractory, in cutting tools, and to protect thermocouple tubes. It is also acts as an antioxidant in carbon-bonded refractory ceramics, neutron absorber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12045-64-6
Molecular Formula B2Zr
Molecular Weight (g/mol) 112.844
MDL Number MFCD00064648
InChI Key VWZIXVXBCBBRGP-UHFFFAOYSA-N
PubChem CID 9812765
IUPAC Name boron;zirconium
SMILES [B].[B].[Zr]

Specifications

Melting Point 3000°C
Physical Form Powder
Quantity 250 g
Linear Formula ZrB2
UN Number UN3178
Solubility Information It is insoluble in water.
Formula Weight 112.84
Percent Purity 99.5%
Assay (metals basis excluding Hf)
Chemical Name or Material Zirconium boride
Hazard Category Warning
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
EINECSNumber 234-963-5
RTECSNumber ZH7150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.