Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zirconium boride, 99.5% (metals basis excluding Hf), Thermo Scientific Chemicals

Catalog No. AA1214030
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1214030 250 g
AA1214018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1214030 Supplier Thermo Scientific Chemicals Supplier No. 01214030
Add to Cart

Zirconium boride is use to improve resistance in zirconia-based, carbon-bonded refractories in contact with ferrous melts. Used in nuclear applications, aerospace refractory, in cutting tools and to protect thermocouple tubes. It also used in use of its relatively high conductivity, especially for a ceramic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12045-64-6
Molecular Formula B2Zr
Molecular Weight (g/mol) 112.844
MDL Number MFCD00064648
InChI Key VWZIXVXBCBBRGP-UHFFFAOYSA-N
Synonym Zirconium diboride
PubChem CID 9812765
IUPAC Name boron;zirconium
SMILES [B].[B].[Zr]

Specifications

Melting Point 3000°C
Physical Form Powder
Quantity 250 g
UN Number UN3178
Solubility Information Insoluble in water.
Formula Weight 112.84
Percent Purity 99.5%
Packaging Plastic bottle
Chemical Name or Material Zirconium boride
EINECSNumber 234-963-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.