Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zirconium carbide, 99.5% (metals basis excluding Hf), Hf< 200ppm, Thermo Scientific Chemicals

Catalog No. AA3580830
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3580830 250 g
AA3580818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3580830 Supplier Thermo Scientific Chemicals Supplier No. 03580830
Add to Cart

missing translation for 'validMainframeMsg'

Zirconium carbide commercially used in tool bits for cutting tools. It is used as an abrasive, in cladding, in cermets and incandescent filaments. It finds application in the coating of uranium dioxide and thorium dioxide particles of nuclear fuel as well as in thermo-photovoltaic radiators and field emitter tips and arrays. It is involved in the usage as refractory coatings in nuclear reactors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12070-14-3
Molecular Formula CZr
Molecular Weight (g/mol) 103.24
MDL Number MFCD00049640
InChI Key MHMXYYDKIQTOHA-UHFFFAOYSA-N
IUPAC Name methanidylidynezirconiumylium
SMILES [C-]#[Zr+]

Specifications

Melting Point 3540°C
Color Gray
Physical Form Powder
Quantity 250 g
Odor Odorless
Density 6.73 g/mL
UN Number UN3178
Solubility Information Soluble in concentrated sulfuric acid,hydrofluoric acid and nitric acid. Insoluble in water.
Formula Weight 103.23
Percent Purity 99.5%
Assay (metals basis excluding Hf), Hf< 200ppm
Chemical Name or Material Zirconium carbide
Show More Show Less
Hazard Category H228
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
EINECSNumber 235-125-1
RTECSNumber ZH7155000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.