Learn More
ALADDIN SCIENTIFIC CORPORATION Zirconium dioxide, 100g
Catalog No. 5000082574
Encompass_Preferred
Zirconium(IV) oxide is used as an additive in the production of piezoelectric crystals in high-frequency induction coils and in ceramic glazes It finds application as a solid electrolyte in electrochromic devices and precursor to the electroceramic lead zirconate titanate (PZT) as an insulator in transistors as a thermal barrier coating in jet and diesel engines and as a diamond simulant in jewelry It is used in oxygen sensors fuel cell membranes and in orthopaedic implants of hip joints It is coated on the silica gel surface and found to be used in the extraction of chromium from aqueous solutions at a pH of about 3
Catalog No. 50-000-82574 Supplier ALADDIN SCIENTIFIC CORPORATION Supplier No. Z104402100G
Zirconium(IV) oxide is used as an additive in the production of piezoelectric crystals in high-frequency induction coils and in ceramic glazes It finds application as a solid electrolyte in electrochromic devices and precursor to the electroceramic lead zirconate titanate (PZT) as an insulator in transistors as a thermal barrier coating in jet and diesel engines and as a diamond simulant in jewelry It is used in oxygen sensors fuel cell membranes and in orthopaedic implants of hip joints It is coated on the silica gel surface and found to be used in the extraction of chromium from aqueous solutions at a pH of about 3