Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Zirconium(II) Hydride

Catalog No. 208558100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-855-8100G 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 20-855-8100G Supplier Sigma Aldrich Supplier No. 208558100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.Zirconium(II) hydride (ZrH2) can be used for the preparation of zirconium diboride (ZrB2) by the reactive plasma sintering process. It can be used as a neutron shielding material, which finds potential usage in hydrogen storage.

Specifications

CAS 13826-86-3
Density 1.268 g/mL at 25°C
Molecular Formula BF4NO2
Linear Formula NO2BF4
MDL Number MFCD00011432
Quantity 200 mL
Synonym Zirconium dihydride
Molecular Weight (g/mol) 132.81 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.