Learn More
Strem, An Ascensus Company CAS# 17501-44-9. 25g. Zirconium(IV) acetylacetonate, min. 97%. MFCD00000036
SDP
Catalog No. 5090115250
Encompass
CAS# 17501-44-9. 25g. Zirconium(IV) acetylacetonate, min. 97%. MFCD00000036. Molecular Weight: 487.66. Molecular Formula: Zr(CH3COCHCOCH3)4. Color/form: white pwdr. Strem# 93-4005. http://www.strem.com/catalog/v/93-4005/
Catalog No. 50-901-15250 Supplier Strem, An Ascensus Company Supplier No. 93400525G
CAS# 17501-44-9. 25g. Zirconium(IV) acetylacetonate, min. 97%. MFCD00000036. Molecular Weight: 487.66. Molecular Formula: Zr(CH3COCHCOCH3)4. Color/form: white pwdr. Strem# 93-4005. http://www.strem.com/catalog/v/93-4005/