Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Zirconium(IV) butoxide solution

Catalog No. 333948100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-394-8100ML 100 mL
33-394-82L 2 L
33-394-8500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 33-394-8100ML Supplier Sigma Aldrich Supplier No. 333948100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used in an unconventional method to prepare a gel of CeO2-stabilized zirconia polycrystals that have promising applications as protective coatings of steel and ferrous alloys.

Specifications

CAS 1071-76-7
Density 1.049 g/mL (at 25°C)
Molecular Formula C16H36O4Zr
Refractive Index n20/D 1.466
Linear Formula Zr(OC4H9)4
MDL Number MFCD00048765
Quantity 100 mL
Synonym Tetrabutyl zirconate solution
Molecular Weight (g/mol) 383.68
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.