Learn More
Strem, An Ascensus Company CAS# 12671-00-0. 2kg. Zirconium(IV) carbonate, basic hydrate. MFCD00172265
SDP
Supplier:  Strem, An Ascensus Company 9340072000G
Encompass
CAS# 12671-00-0. 2kg. Zirconium(IV) carbonate, basic hydrate. MFCD00172265. Molecular Weight: 431.68. Molecular Formula: [Zr3(CO3)O5]H2O. Color/form: white pwdr. Strem# 93-4007. http://www.strem.com/catalog/v/93-4007/
Catalog No. 50-901-15255