Learn More
Strem, An Ascensus Company CAS# 10026-11-6. 50g. Zirconium(IV) chloride (99.5+%-Zr). MFCD00011306
SDP
Catalog No. 5090115287
Encompass
CAS# 10026-11-6. 50g. Zirconium(IV) chloride (99.5+%-Zr). MFCD00011306. Molecular Weight: 233.03. Molecular Formula: ZrCl4. Color/form: white pwdr. Strem# 93-4045. http://www.strem.com/catalog/v/93-4045/
Catalog No. 50-901-15287 Supplier Strem, An Ascensus Company Supplier No. 93404550G
CAS# 10026-11-6. 50g. Zirconium(IV) chloride (99.5+%-Zr). MFCD00011306. Molecular Weight: 233.03. Molecular Formula: ZrCl4. Color/form: white pwdr. Strem# 93-4045. http://www.strem.com/catalog/v/93-4045/