Learn More
Strem, An Ascensus Company CAS# 14717-56-7. 10g. Zirconium(IV) i-propoxide (isopropanol adduct) (99.9%-Zr). MFCD00078045
SDP
Supplier:  Strem, An Ascensus Company 40401610G
Encompass
CAS# 14717-56-7. 10g. Zirconium(IV) i-propoxide (isopropanol adduct) (99.9%-Zr). MFCD00078045. Molecular Weight: 327.56 (387.67). Molecular Formula: Zr(OC3H7)4∙C3H7OH. Color/form: white xtl. Strem# 40-4016. http://www.strem.com/catalog/v/40-4016/
Catalog No. 50-901-11617