Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Zirconium(IV) oxide-yttria stabilized

Catalog No. 464228500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-422-8500G 500 g
46-422-8100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 46-422-8500G Supplier Sigma Aldrich Supplier No. 464228500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Zirconium(IV) oxide-yttria stabilized

Specifications

CAS 114168-16-0
Melting Point >2,600°C
Molecular Formula O5Y2Zr
Linear Formula ZrO2 -+ Y2O3
MDL Number MFCD00145546
Quantity 500 g
Synonym YSZ; Yttria stabilized zirconia; Yttrium stabilized zirconium oxide
Molecular Weight (g/mol) 349.03
Percent Purity 99.9% trace metals basis (purity excludes ∼2%HfO2)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.