Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Zirconium(IV) oxynitrate hydrate

Catalog No. 2434935G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-349-35G 5 g
24-349-3500G 500 g
24-349-3100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 24-349-35G Supplier Sigma Aldrich Supplier No. 2434935G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Zirconium(IV) oxynitrate hydrate is used to prepare CeO2-ZrO2 mixed oxide catalyst supports. It may be used as zirconium precursor in the preparation of;Pb(Zr0.50Ti0.50)O3 ferroelectric thin films, silica- and alumina-doped nanocrystalline ceria/zirconia (Ce0.5Zr0.5O2).

Specifications

CAS 14985-18-3
Quantity 5 g
Molecular Formula N2O7Zr -+ xH2O
Linear Formula ZrO(NO3)2 -+ xH2O
MDL Number MFCD00149896
Synonym Zirconyl nitrate hydrate
Molecular Weight (g/mol) 231.23
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.