Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Zirconium(IV) propoxide solution

Catalog No. 333972500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-397-2500ML 500 mL
33-397-2100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 33-397-2500ML Supplier Sigma Aldrich Supplier No. 333972500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Zirconium(IV) propoxide solution is a metal alkoxide solution that is highly reactive with water. It can be used as a precursor material for the synthesis of zirconium oxide thin films by atmospheric pressure electrospray. Zirconium(IV) propoxide can also be used in the preparation of stable sols for potential usage in optical, electronic and magnetic applications.

Specifications

CAS 23519-77-9
Density 1.044 g/mL (at 25°C)
Boiling Point 208°C (0.1 mmHg)
Molecular Formula C12H28O4Zr
Linear Formula Zr(OCH2CH2CH3)4
Refractive Index n20/D 1.451
Quantity 500 mL
MDL Number MFCD00015307
Synonym Tetrapropyl zirconate
Molecular Weight (g/mol) 327.57
Biological Activity Central nervous system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.