Learn More
Strem, An Ascensus Company CAS# 12039-15-5. 10g. Zirconium(IV) sulfide, 99%. MFCD00054116
SDP
Catalog No. 5090115276
Encompass
CAS# 12039-15-5. 10g. Zirconium(IV) sulfide, 99%. MFCD00054116. Molecular Weight: 155.35. Molecular Formula: ZrS2. Color/form: -200 mesh reddish-brown pwdr. Strem# 93-4033. http://www.strem.com/catalog/v/93-4033/
Catalog No. 50-901-15276 Supplier Strem, An Ascensus Company Supplier No. 93403310G
CAS# 12039-15-5. 10g. Zirconium(IV) sulfide, 99%. MFCD00054116. Molecular Weight: 155.35. Molecular Formula: ZrS2. Color/form: -200 mesh reddish-brown pwdr. Strem# 93-4033. http://www.strem.com/catalog/v/93-4033/