Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Zirconium(IV) tert-butoxide

Catalog No. 5600305G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-003-05G 5 g
56-003-025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 56-003-05G Supplier Sigma Aldrich Supplier No. 5600305G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Zirconium(IV) tert-butoxide may be used to synthesize chiral N-tert-butanesulfinyl trifluoromethyl ketimines. Zirconia coatings on stainless steel substrates for optical applications can be prepared from zirconium (IV) tert-butoxide precursor by sol gel method.

Specifications

Density 0.985 g/mL (at 25°C)
Boiling Point 81°C (3 mmHg)
Molecular Formula C16H36O4Zr
Refractive Index n20/D 1.424 (literature)
Linear Formula Zr[OC(CH3)3]4
Quantity 5 g
MDL Number MFCD00075085
Synonym Tetra-tert-butoxyzirconium; ZTB; Zirconium tert-butanolate; Tetra-tert-butyl zirconate
Molecular Weight (g/mol) 383.68
Percent Purity 99.999% trace metals basis
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.