Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zirconium(IV) tert-butoxide, 97+%, Thermo Scientific Chemicals

Catalog No. AA4258306
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4258306 5 g
AA4258303 1 g
AA4258314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4258306 Supplier Thermo Scientific Chemicals Supplier No. 04258306
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2081-12-1
Molecular Formula C16H40O4Zr
Molecular Weight (g/mol) 387.72
MDL Number MFCD00075085
InChI Key WRMYASGYMABHCC-UHFFFAOYSA-N
Synonym zirconium iv tert-butoxide, 2-methylpropan-2-ol; zirconium
PubChem CID 25203936
IUPAC Name 2-methylpropan-2-ol;zirconium
SMILES CC(C)(C)O.CC(C)(C)O.CC(C)(C)O.CC(C)(C)O.[Zr]

Specifications

Color Yellow
Density 0.985 g/mL
Boiling Point 90°C (5 mmHg)
Flash Point 85°C (185°F)
Linear Formula Zr[OC(CH3)3]4
Quantity 5 g
Beilstein 3681870
Sensitivity Light and moisture sensitive
Formula Weight 383.68
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material Zirconium (IV) tert-butoxide
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.