Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zirconium oxide, catalyst support, monoclinic phase, Thermo Scientific Chemicals

Catalog No. AA43815A4
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43815A4 2.5 kg
AA4381522 100 g
AA4381536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA43815A4 Supplier Thermo Scientific Chemicals Supplier No. 043815A4
Only null left
Add to Cart

Typical properties S.A. 90m2/g, median pore diameter 80/400Å, bimodal total pore volume 0.31cc/g, packing density 1.181g/L

Zirconium oxide is used as a catalyst carrier, diamond simulant, refractory material, pigment, abrasives and solid electrolyte in electrochromic devices. It acts as a precursor to the electroceramic lead zirconate titanate. It is also used in insulation, oxygen sensors, enamels and fuel cell. It finds application to make ceramic knives due to its hardness nature. It is utilized for dental restorations like crowns.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00011310

Specifications

Physical Form Pellets
Quantity 2.5 kg
Solubility Information Soluble in hot-concentrated hydrogen fluoride and sulfuric acid. Insoluble in water.
Chemical Name or Material Zirconium oxide, catalyst support, 1/8 in. Pellets
Hazard Category H500
EINECSNumber 215-227-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.