Learn More
Strem, An Ascensus Company CAS# 7440-67-7. 100g. Zirconium powder (99.5%). MFCD00011303
SDP
Catalog No. 5090115291
Encompass
CAS# 7440-67-7. 100g. Zirconium powder (99.5%). MFCD00011303. Molecular Weight: 91.22. Molecular Formula: Zr. Color/form: -50 mesh pwdr. Strem# 93-4047. http://www.strem.com/catalog/v/93-4047/
Catalog No. 50-901-15291 Supplier Strem, An Ascensus Company Supplier No. 934047100G
CAS# 7440-67-7. 100g. Zirconium powder (99.5%). MFCD00011303. Molecular Weight: 91.22. Molecular Formula: Zr. Color/form: -50 mesh pwdr. Strem# 93-4047. http://www.strem.com/catalog/v/93-4047/