Learn More
Strem, An Ascensus Company CAS# 7440-67-7. 5cm. Zirconium rod (99.2+%). MFCD00011303
SDP
Catalog No. 5090115293
Encompass
CAS# 7440-67-7. 5cm. Zirconium rod (99.2+%). MFCD00011303. Molecular Weight: 91.22. Molecular Formula: Zr. Color/form: 12.7 mm dia. (~8.2g/cm) Strem# 93-4049. http://www.strem.com/catalog/v/93-4049/
Catalog No. 50-901-15293 Supplier Strem, An Ascensus Company Supplier No. 9340495CM
CAS# 7440-67-7. 5cm. Zirconium rod (99.2+%). MFCD00011303. Molecular Weight: 91.22. Molecular Formula: Zr. Color/form: 12.7 mm dia. (~8.2g/cm) Strem# 93-4049. http://www.strem.com/catalog/v/93-4049/