Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zirconium silicide, 99.5% (metals basis excluding Hf), Hf 4% max, Thermo Scientific Chemicals

Catalog No. p-7050257
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3632214 25 g
AA3632222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3632214 Supplier Thermo Scientific Chemicals Supplier No. 03632214
Only null left
Add to Cart

Zirconium silicate is used for manufacturing refractory materials. It is also used in production of some ceramics, enamels, and ceramic glazes. In enamels and glazes it serves as an opacifier. zirconium silicate is as beads for milling and grinding.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12039-90-6
Molecular Formula H4Si2Zr
Molecular Weight (g/mol) 151.43
MDL Number MFCD00151547
InChI Key ZSONHIWWHDAZQO-UHFFFAOYSA-N
Synonym zirconium silicide, zirconium silicide zrsi2, zirconium silicide, zrsi2, $l^ 5-silanylidenezirconiumylidene-$l^ 5-silane
PubChem CID 6336892
IUPAC Name bis($l^{2}-silanylidene)zirconium
SMILES [SiH2]=[Zr]=[SiH2]

Specifications

Color Gray
Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water. It is insoluble in acids,alkali and aqua regia.
Formula Weight 147.39
Percent Purity 99.5%
Odor Odorless
Density 4.88 g/mL
Assay (metals basis excluding Hf), Hf 4%
Chemical Name or Material Zirconium silicide
EINECSNumber 234-911-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.