Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zirconium titanium oxide, 99.5% (metals basis excluding Hf), Hf <75ppm, Thermo Scientific Chemicals

Catalog No. AA3949014
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3949014 25 g
AA3949022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3949014 Supplier Thermo Scientific Chemicals Supplier No. 03949014
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Suitable for glass, optic and ceramic applications, advanced electronics and in light weight structural components in aerospace and electrochemical applications such as fuel cells in which they exhibit ionic conductivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12036-70-3
MDL Number MFCD00210648
Synonym zirconium titanium oxide, tetrahydrate titanium zirconium

Specifications

Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 203.11
Percent Purity 99.5%
Odor Odorless
Assay (metals basis excluding Hf), Hf <75ppm
Chemical Name or Material Zirconium titanium oxide
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.