Learn More
Next Advance Inc Zirconium Oxide Beads 2.0 mm RNase Free, 4 mL
SDP
Supplier:  Next Advance Inc ZROB20RNA
Encompass
2.0 mm Zirconium Oxide beads, RNase free, density is 5.5 g/cc, 4 mL package.
Catalog No. NC1113340