Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zirconocene Chloride Hydride 96.0+%, TCI America™
SDP

Catalog No. Z001025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z001025G 25 g
Z00101G 1 g
Z00105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. Z001025G Supplier TCI America Supplier No. Z001025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 37342-97-5
Molecular Formula C10H11ClZr-2
Molecular Weight (g/mol) 257.872
MDL Number MFCD02089401
InChI Key VPQMECJLNPUNGN-UHFFFAOYSA-M
Synonym zirconocene chloride hydride
PubChem CID 131718033
IUPAC Name cyclopenta-1,3-diene;hydride;zirconium(2+);chloride
SMILES [H-].[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Zr+2]

Specifications

Quantity 25 g
UN Number 3131
Formula Weight 257.87
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Zirconocene Chloride Hydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.