Learn More
Medchemexpress LLC HY-109142 10mg Medchemexpress, Ziresovir CAS:1422500-60-4 Purity:>98%
SDP
Catalog No. 501968687
Encompass_Preferred
Medchemexpress, HY-109142 10mg Ziresovir CAS:1422500-60-4 Ziresovir (AK0529;RO-0529) is a potent, selective, and orally bioavailable respiratory syncytial virus (RSV) fusion (F) protein (RSV F) protein inhibitor. Ziresovir shows anti-RSV activity (EC50=3 nM) and highlights pharmacokinetics in animal species. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8687 Supplier Medchemexpress LLC Supplier No. HY10914210MG
Medchemexpress, HY-109142 10mg Ziresovir CAS:1422500-60-4 Ziresovir (AK0529;RO-0529) is a potent, selective, and orally bioavailable respiratory syncytial virus (RSV) fusion (F) protein (RSV F) protein inhibitor. Ziresovir shows anti-RSV activity (EC50=3 nM) and highlights pharmacokinetics in animal species. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.