Learn More
Medchemexpress LLC Ziresovir | 1422500-60-4 | 95.3% | 439.53 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY109142100MG
Encompass_Preferred
Ziresovir is a potent, selective, and orally bioavailable respiratory syncytial virus (RSV) fusion (F) protein inhibitor. It demonstrates anti-RSV activity (EC50=3 nM) and favorable pharmacokinetics in animal species.
  • Potent and selective RSV F protein inhibitor.
  • Orally bioavailable.
  • Demonstrates anti-RSV activity with an EC50 of 3 nM.
  • Favorable pharmacokinetics in animal species.
Catalog No. 50-003-45460