Learn More
Coast Zithion-X AAA Rechargeable Batteries
Catalog No. 17100474
Change view
Click to view available options
Quantity:
12/Pk.
4/Pk.
8/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-100-474 12/Pk.
17-100-475 8/Pk.
17-100-476 4/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-100-474 Supplier Coast Supplier No. 30987
Add to Cart
Add to Cart
Pack power with purpose. ZITHION-X™ rechargeable lithium batteries change the game in responsible, portable power technology.
  • COAST ZITHION-X™ rechargeable lithium batteries change the game in responsible, portable power technology.
  • Advanced engineering provides a comprehensive upgrade compared with other sources of battery power by delivering long-lasting run times for a clear performance advantage over standard rechargeable lithium and NiMH batteries.
  • To mimic the broad compatibility of alkaline batteries, proprietary COAST ZX Optimized Voltage™ allows ZITHION-X batteries to modulate their power supply up or down based on the need and prevent flickering, overheating, and damage to products.
  • The leak-resistant battery casing prevents corrosion during use, matching performance with protection.
  • Connect conveniently via USB-C to recharge quickly (in 2.5 hours or less for AA and 1.25 hours or less for AAA) and forget about special charging cables and cradles. Every ZITHION-X can be recharged at least 1000 times, resulting in significant cost-savings compared with single-use disposable batteries while keeping hundreds of batteries out of the landfill.
Specifications
Product Type Rechargeable Battery
Battery Type AAA
Material Lithium-ion
Cell Type Spring Terminal
Quantity 12/Pk.
Voltage 1.5 V