Learn More
COAST Cutlery Zithion-X USB-C Ported Battery
Catalog No. 19030059
Change view
Click to view available options
Battery Type:
AA
AAA
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Battery Type
19-030-059 AA
19-030-060 AAA
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-030-059 Supplier COAST Cutlery Supplier No. 31007
Add to Cart
Edge
Add to Cart
Pack power with purpose. COAST ZITHION-X™ rechargeable lithium batteries change the game in responsible, portable power technology.
  • Battery Charging Level Indicator - Check the status of your batteries charge level with our built-in charge level indicator. (Solid red) when charging, (solid green) when fully charged.
  • Built-In USB-C Port - With a built-in USB-C port, your battery can be charged using a USB-C cord. Some COAST models can be charged both inside or outside the light.
  • Overcharge/Discharge Protection Circuits - Over charging and improperly discharging lithium batteries can cause a safety concern and hazardous risk to the user and it's surroundings. COAST batteries are built with overcharge/discharge protection, ensuring you and your battery are protected.
Specifications
Product Type Replacement Battery
Battery Type AA
Material Lithium-ion
Quantity 4/Pk.
Voltage 1.5 V