Learn More
Cayman Chemical ZIyuglycosIde I 10mg
SDP
Catalog No. 502917378
Encompass_Preferred
A triterpenoid saponin with diverse biological activities; increases the synthesis of type I collagen of CCD-1064Sk human fibroblast cells; inhibits the proliferation of MDA-MB-231 breast cancer cells (IC50 = 13.96 µM); induces apoptosis in MDA-MB-231 cells and also increases levels of p53 and p21WAF1 in MDA-MB-231 cells
Catalog No. 50-291-7378 Supplier Cayman Chemical Supplier No. 3732010 MG
A triterpenoid saponin with diverse biological activities; increases the synthesis of type I collagen of CCD-1064Sk human fibroblast cells; inhibits the proliferation of MDA-MB-231 breast cancer cells (IC50 = 13.96 µM); induces apoptosis in MDA-MB-231 cells and also increases levels of p53 and p21WAF1 in MDA-MB-231 cells