Learn More
Medchemexpress LLC HY-N0331 5mg Medchemexpress, Ziyuglycoside I CAS:35286-58-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN03315MG
Encompass_Preferred
Medchemexpress, HY-N0331 5mg Ziyuglycoside I CAS:35286-58-9 Ziyuglycoside I isolated from S. officinalis root, has anti-wrinkle activity, and increases the expression of type I collagen. Ziyuglycoside I could be used as an active ingredient for cosmetics.Ziyuglycoside I triggers cell cycle arrest and apoptosis mediated by p53, it can be a potential drug candidate for treating triple-negative breast cancer (TNBC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0253