Learn More
Cayman Chemical ZIyuglycosIde II 10mg
SDP
Catalog No. 502942814
Encompass_Preferred
A glycoside with diverse biological activities; cytotoxic to SMMC-7721, PC-9, HCT116, MCF-7, and MDA-MB-231 cancer cells at 30 and 90 UG/ml; increases the number of white blood cells in peripheral blood and the number of HSPCs in a mouse model of cyclophosphamide-induced leukopenia at 6, 12, and 18 mg/kg; reduces small intestinal epithelial cell apoptosis and IL-1β, IL-6, and TNF-α levels, as well as improves diarrhea symptoms in a mouse model of rotavirus-induced diarrhea
Catalog No. 50-294-2814 Supplier Cayman Chemical Supplier No. 4083110 MG
A glycoside with diverse biological activities; cytotoxic to SMMC-7721, PC-9, HCT116, MCF-7, and MDA-MB-231 cancer cells at 30 and 90 UG/ml; increases the number of white blood cells in peripheral blood and the number of HSPCs in a mouse model of cyclophosphamide-induced leukopenia at 6, 12, and 18 mg/kg; reduces small intestinal epithelial cell apoptosis and IL-1β, IL-6, and TNF-α levels, as well as improves diarrhea symptoms in a mouse model of rotavirus-induced diarrhea