Learn More
Cayman Chemical ZK 118182 Isopropyl esTR 100ug
SDP
Catalog No. 502953624
Encompass_Preferred
A prodrug formulation of a potent DP-receptor agonist that is designed to enhance corneal absorption; lowers monkey and rabbit IOP 46% and 20%, respectively, two hours after dosing at 0.03 μg demonstrating a much more potent effect compared to the free acid
Catalog No. 50-295-3624 Supplier Cayman Chemical Supplier No. 13673100 UG
A prodrug formulation of a potent DP-receptor agonist that is designed to enhance corneal absorption; lowers monkey and rabbit IOP 46% and 20%, respectively, two hours after dosing at 0.03 μg demonstrating a much more potent effect compared to the free acid